Anti-MYCN Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP7395B
ABGEAP7395BEA 345 GBP
ABGEAP7395B
Anti-MYCN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MYCN (v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR