Anti-ERMN Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP4892C
ABGEAP4892CEA 345 GBP
ABGEAP4892C
Anti-ERMN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ERMN (ermin, ERM-like protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR