Anti-ZBTB17 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP18958A
ABGEAP18958AEA 345 GBP
ABGEAP18958A
Anti-ZBTB17 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZBTB17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR