Anti-RAB22A Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP18867B
ABGEAP18867BEA 345 GBP
ABGEAP18867B
Anti-RAB22A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB22A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR