Anti-MNX1 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP16474C
ABGEAP16474CEA 345 GBP
ABGEAP16474C
Anti-MNX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MNX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR