Anti-TNNI2 Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP16030A
ABGEAP16030AEA 345 GBP
ABGEAP16030A
Anti-TNNI2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TNNI2 (troponin I type 2 (skeletal, fast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR