Anti-RAD9A Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP1318A
ABGEAP1318AEA 345 GBP
ABGEAP1318A
Anti-RAD9A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: BH3 domain
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR