Anti-TTC9B Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP11588B
ABGEAP11588BEA 345 GBP
ABGEAP11588B
Anti-TTC9B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TTC9B (tetratricopeptide repeat domain 9B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

SPECIFICATIONS

Antigen symbol TTC9B
Antigen name Tetratricopeptide Repeat Doman 9B
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Mouse
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Epitope C-terminal
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR