Anti-SAR1B Rabbit Polyclonal Antibody

Supplier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP10397C
ABGEAP10397CEA 345 GBP
ABGEAP10397C
Anti-SAR1B Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SAR1B (secretion associated, Ras related GTPase 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR