2-Chlorocinnamic acid predominantly trans ≥99%

Supplier: ABCR
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

AB177468.0100
ABCRAB177468.0100EA 0 GBP
ABCRAB177468.0100
2-Chlorocinnamic acid predominantly trans ≥99%
2-Chlorocinnamic acid

Purity: 99% CAS No.: 3752-25-8 Molecular Formula: C9H7ClO2 Formula Weight: 182.61 Melting Point: 209-212° Beilstein Reference: 1365198

Formula: C₉H₇ClO₂
MW: 182.61 g/mol
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 209…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00004372
CAS Number: 3752-25-8
EINECS: 223-154-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR