Sodium orthosilicate

Supplier: abcr
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Tetrasodium orthosilicate, Sodium silicate (ortho)

AB122045.1000
ABCRAB122045.1000EA 0 GBP
ABCRAB122045.1000
Sodium orthosilicate
tetra-Sodium orthosilicate
Formula: Na₄O₄Si
Melting Pt: 1018 °C
Density: 2.5 g/cm³ (26 °C)
MDL Number: MFCD00014252
CAS Number: 13472-30-5
EINECS: 236-741-3
UN: 1759
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR