Anti-CD3 Mouse Monoclonal Antibody [clone: FN-18]

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB25109-100
ABCAAB25109-100EA 856 GBP
ABCAAB25109-100
Anti-CD3 Mouse Monoclonal Antibody [clone: FN-18]
Antibodies
Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of

More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD3
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone FN-18
Reactivity
Host Mouse
Isotype IgG1
Flow cytometry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR