Anti-SRM Rabbit Polyclonal Antibody

Supplier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB111884-50
ABCAAB111884-50EA 395 GBP
ABCAAB111884-50
Anti-SRM Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SRM (spermidine synthase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR