Biotinyl tyramide for signal amplification

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Biotin-Phenol, N-(4-Hydroxyphenethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

45350
AATB45350EA 127 GBP
AATB45350
Biotinyl tyramide for signal amplification
Biotinyl tyramide

For many immunohistochemical (IHC) applications, the traditional enzymatic amplification procedures are sufficient for achieving adequate antigen detection.


  • Signal amplification
  • Highest IHC detection of all TSA reagents
  • Suitable for multiple applications

Formula: C₁₈H₂₅N₃O₃S
MW: 363.5 g/mol
MDL Number: MFCD27953026
CAS Number: 41994-02-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0bd24a3d8b1a40ebaf130a85dafb0af1fba1cc45&vid=7fc369374bfe081348b99b7e58f85155454dcce2&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR