Thioflavin T succinimidyl ester ≥90%
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
23065
AATB23065EA 271 GBP
AATB23065
Thioflavin T succinimidyl ester ≥90%
Thioflavin T succinimidyl ester
Thioflavin T (ThT), also called Basic Yellow 1 or CI 49005, is a benzothiazole salt.
  • Essential Thioflavin T Reagent
  • Useful In Alzheimer's Research
Caution: For research use only.