Congo red ≥95%, Ultra Pure Grade

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

23056
AATB23056EA 106 GBP
AATB23056
Congo red ≥95%, Ultra Pure Grade
Congo Red

Congo red (CR) is the sodium salt of benzidinediazo-bis-1-naphtylamine-4-sulfonic acid.

Formula: C₃₂H₂₂N₆Na₂O₆S₂
MDL Number: MFCD00004028
CAS Number: 573-58-0
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR