Cell Meter™ Colorimetric MTS Cell Proliferation Kit
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22766 22767
AATB22766EA 382 GBP
AATB22766 AATB22767
Cell Meter™ Colorimetric MTS Cell Proliferation Kit
Assays Cellular Assays
Designed for consistent and dependable results. The Cell Meter™ Colorimetric MTS Cell Proliferation Kit is a quick and reliable method for quantifying viable cells in proliferation and cytotoxicity assays.
  • Can be applied directly to cell culture media, bypassing the intermediate steps required by traditional MTT assays
  • Absorbance measured at 490 to 500 nm
  • Eliminates the need for washing or solubilisation steps
The Cell Meter™ Colorimetric MTS Cell Proliferation Kit is useful for evaluating cell proliferation in response to various stimuli such as growth factors, cytokines, mitogens and nutrients
Delivery information: Shipped with dry ice
Caution: Research use only.