MitoLite™ Deep Red FX660
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22678
AATB22678EA 135 GBP
AATB22678
MitoLite™ Deep Red FX660
Assays Cellular Assays
MitoLite™ deep red is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Caution: For research use only.