Cell Navigator® Live Cell Endoplasmic Reticulum (ER) Staining Kit *Red Fluorescence*
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22636
AATB22636EA 406 GBP
AATB22636
Cell Navigator® Live Cell Endoplasmic Reticulum (ER) Staining Kit *Red Fluorescence*
Assays Cellular Assays
The endoplasmic reticulum (ER) is a type of organelle in the cells of eukaryotic organisms that forms an interconnected network of flattened, membrane-enclosed sacs or tube-like structures known as cisternae.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.