Rhodamine B hexyl ester perchlorate
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22211
AATB22211EA 106 GBP
AATB22211
Rhodamine B hexyl ester perchlorate
Rhodamine B hexyl ester perchlorate
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.
  • Selective for mitochondria
  • Easily-detectable signal
  • Multiple applications
Caution: For research use only.