Dextran, M.W. 150,000 TRITC-labeled

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

21710
AATB21710EA 258 GBP
AATB21710
Dextran, M.W. 150,000 TRITC-labeled
Dextran

TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.


  • Well-retained in cells
  • 150000 mw
  • Multiple applications


From Eukaryotic cells.


Caution: For research use only.

Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR