QXY7 NHS ester [equivalent to QSY-7 NHS ester]
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2038
AATB2038EA 250 GBP
AATB2038
QXY7 NHS ester [equivalent to QSY-7 NHS ester]
QXY7 NHS ester
QXY7 is the same molecule to QSY-7 acid (ThermoFisher).
  • Stable for 12 months
Quality tested in-house
Caution: For Research Use Only