ICG-Sulpho-OSu ≥80%
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
180
AATB180EA 406 GBP
AATB180
ICG-Sulpho-OSu ≥80%
ICG-Sulpho-OSu
Indocyanine green (ICG) is a cyanine dye used in medical diagnostics.
  • Dependable in vivo imaging applications
  • Infrared absorption wavelength
  • Ideal for bioconjugation
Caution: For research use only.
CAS Number: 190714-28-4