TAQuest™ qPCR Master Mix for TaqMan Probes *Low ROX*
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17284 17285
AATB17284EA 122 GBP
AATB17284 AATB17285
TAQuest™ qPCR Master Mix for TaqMan Probes *Low ROX*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
TAQuest™ qPCR Master mix for TaqMan probes is a ready-to-use 2X solution optimised for qPCR and two-step RT-qPCR compatible with TaqMan gene expression assays.
  • Simple and robust protocol
  • The optimised composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
TAQuest™ qPCR Master mix for TaqMan probes has been designed to be used for duplex reactions using internal positive controls.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimised light exposure.