TAQuest™ qPCR Master Mix with Helixyte™ Green *No ROX*
Supplier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17270 17271
AATB17270EA 245 GBP
AATB17270 AATB17271
TAQuest™ qPCR Master Mix with Helixyte™ Green *No ROX*
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
TAQuest™ qPCR Master mix with Helixyte™ green is a ready-to-use 2X solution optimised for qPCR and two-step RT-qPCR.
  • Simple and robust protocol
  • The optimised composition ensures PCR specificity and sensitivity with all sample types such as genomic, plasmid, viral and cDNA templates
The Helixyte green intercalating dye allows rapid DNA detection and analysis without using sequence-specific probes.
Caution: Master mix is stable for 12 months when stored frozen (<−15 °C) with minimised light exposure.