Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 860)

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

116100R0 116100R1
AATB116100R0EA 328 GBP
AATB116100R0 AATB116100R1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 860)
Antibodies
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 860
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR