Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (APC/Cy7)

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

101611D0 101611D1 101611D2
AATB101611D0EA 282 GBP
AATB101611D0 AATB101611D1 AATB101611D2
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (APC/Cy7)
Antibodies
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: APC/Cy7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR