Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 500)

Supplier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10041100 10041101
AATB10041100EA 328 GBP
AATB10041100 AATB10041101
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4] (mFluor™ Violet 500)
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Affinity purified
  • Easily reconstituted

Caution: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: mFluor™ Violet 500
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR