Tricaine methanesulphonate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A5040-100G A5040-250G A5040-25G
A5040-100GEA 343 GBP
A5040-100G A5040-250G A5040-25G
Tricaine methanesulphonate, Supelco®
Tricaine methanesulphonate
Formula: C₁₀H₁₅NO₅S
MDL Number: MFCD00013176
CAS Number: 886-86-2
EINECS: 212-956-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4922b270808efb4cd4796a992e52d11b681edb59&vid=5939f58e3f4d09c0b8c2fed5d887d62d9d603e93&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6852165981703cc677bb882168b1c43d47dc7cd0&vid=ff3fba20032b3c8c90e8697d8ab761f11536f2ed&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10f5602a3920810e00d619c66075a20b10cedab4&vid=d26a43c1e111342685b80246f2e510c9cd1ddb9f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR