Acrylamide : Bisacrylamide (37.5:1) 40% aqueous solution

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Acrylamide- N,N'-Methylenebisacrylamide, Acrylamide-Bis

A4989.0500
A4989.0500EA 138 GBP
A4989.0500
Acrylamide : Bisacrylamide (37.5:1) 40% aqueous solution
Acrylamide-Bis (32:1)
Storage Temperature: Ambient
MDL Number: MFCD00080848
UN: 3426
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR