Buffer, triethylammonium acetate, 1 M solution, pH 7,0

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A3846.0500 A3846.1000
A3846.0500EA 142 GBP
A3846.0500 A3846.1000
Buffer, triethylammonium acetate, 1 M solution, pH 7,0
Buffers Biological Buffers

Used for the purification of oligonucleotides.


CAS No.: 5204-74-0
Molecular Formula: C₆H₁₅N · CH₃COOH
Formula Weight: 161,24 g/mol
Density: 1,001 (at 20 °C)
Physical Form: Liquid
Non-volatile Matter: Max. 0,01%
Storage Temperature: Ambient

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR