L(+)-Asparagine monohydrate

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A3721.1000
A3721.1000EA 252 GBP
A3721.1000
L(+)-Asparagine monohydrate
L(+)-Asparagine monohydrate
Formula: C₄H₈N₂O₃·1H₂O
Melting Pt: 215…217 °C
Density: 1.567 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151038
CAS Number: 5794-13-8
EINECS: 200-735-9
Merck Index: 14,00837

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR