Sodium chloride ≥98.5% for molecular biology

Supplier: PanReac AppliChem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Saline, NaCl, Sea salt, Halite

A2942.1000 A2942.5000
A2942.1000EA 52.5 GBP
A2942.1000 A2942.5000
Sodium chloride ≥98.5% for molecular biology
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

DNases/RNases/Proteases not detectable
Assay (titr.) min. 99.5 %
pH (5 %; H2O; 20°C) 5.0 - 8.0
Heavy metals (as Pb) max. 0.0005 %
Bromide max. 0.005 %
Iodide max. 0.001 %
Phosphate max. 0.001 %
Sulfate max. 0.01 %
Ba max. 0.001 %
Ca max. 0.002 %
Fe max. 0.0001 %
K max. 0.01 %
MG max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR