9-Decen-1-ol ≥90%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Decen-10-ol

A19490.14 A19490.22
A19490.14EA 44.2 GBP
A19490.14 A19490.22
9-Decen-1-ol ≥90%
9-Decen-1-ol

Purity: 90+% CAS No.: 13019-22-2 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 104°(219°F) Refractive Index: 1.4480 Odour: Pleasant Melting Point: -13° Boiling Point: 235° Beilstein Reference: 1750928 MDL No.: MFCD00002992

Formula: C₁₀H₂₀O
Boiling Pt: 235 °C
Melting Pt: –13 °C
Density: 0.847
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002992
CAS Number: 13019-22-2
EINECS: 235-878-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR