N-Cbz-L-serine 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A19050.06 A19050.14 A19050.22
A19050.06EA 22.6 GBP
A19050.06 A19050.14 A19050.22
N-Cbz-L-serine 99%
N-Cbz-L-serine

Purity: 99% CAS No.: 1145-80-8 Molecular Formula: C11H13NO5 Formula Weight: 239.23 Melting Point: 117-120° Beilstein Reference: 2058314 Optical Rotation: +6° (c=7 in acetic acid) MDL No.: MFCD00002662

Formula: C₁₁H₁₃NO₅
Melting Pt: 117…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00002662
CAS Number: 1145-80-8
EINECS: 214-546-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR