(±)-β-Citronellol 95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: DL-Citronellol, (±)-Citronellol, Citronellol, β-Citronellol, 3,7-Dimethyloct-6-en-1-ol

A19016.22 A19016.36
A19016.22EA 22.4 GBP
A19016.22 A19016.36
(±)-β-Citronellol 95%
(±)-β-Citronellol

Purity: 95% CAS No.: 106-22-9 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 98°(208°F) Refractive Index: 1.4575 Odour: Pleasant Melting Point: -20° Boiling Point: 221-222° Beilstein Reference: 1721507 Merck Reference: 14,2330 UN No.: 3082 MDL No.: MFCD00002935

Formula: C₁₀H₂₀O
MW: 156.27 g/mol
Boiling Pt: 221…222 °C
Melting Pt: < –20 °C
Density: 0.858
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002935
CAS Number: 106-22-9
EINECS: 203-375-0
UN: 3082
ADR: 9 III
Merck Index: 13,02354

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR