D-(+)-Tryptophan ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A18426.06 A18426.14 A18426.22
A18426.06EA 25.6 GBP
A18426.06 A18426.14 A18426.22
D-(+)-Tryptophan ≥99%
D(+)-Tryptophan

Purity: 99% CAS No.: 153-94-6 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: 273° dec. Beilstein Reference: 86198

Formula: C₁₁H₁₂N₂O₂
Melting Pt: 273 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064339
CAS Number: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR