2,4,6-Triphenylpyrilium tetrafluoroborate ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2,4,6-Triphenylpyrylium tetrafluoroborate

A18217.06 A18217.14 A18217.22
A18217.06EA 51 GBP
A18217.06 A18217.14 A18217.22
2,4,6-Triphenylpyrilium tetrafluoroborate ≥97%
2,4,6-Triphenylpyrilium tetrafluoroborate

Purity: 97% CAS No.: 448-61-3 Molecular Formula: C23H17BF4O Formula Weight: 396.20 Melting Point: 248-254° Beilstein Reference: 3586533

Formula: C₂₃H₁₇BF₄O
Melting Pt: 248…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00012001
CAS Number: 448-61-3
EINECS: 207-186-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR