Sodium sulphite, anhydrous 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A17933.0C A17933.0E A17933.36
A17933.0CEA 250 GBP
A17933.0C A17933.0E A17933.36
Sodium sulphite, anhydrous 98%
Sodium sulphite

Purity: 98% CAS No.: 7757-83-7 Molecular Formula: Na2SO3 Formula Weight: 126.04 Melting Point: 500° dec. Merck Reference: 14,8682 MDL No.: MFCD00003503

Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: >500 °C (dec.)
Density: 2.633
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 14,8682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR