(1S)-(-)-β-Pinene 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: (-)-Pin-2(10)-ene

A17818.0F A17818.AE A17818.AP
A17818.0FEA 121 GBP
A17818.0F A17818.AE A17818.AP
(1S)-(-)-β-Pinene 99
(1S)-(-)-β-Pinene

Purity: 99% CAS No.: 18172-67-3 Molecular Formula: C10H16 Formula Weight: 136.24 Flash Point: 36°(96°F) Refractive Index: 1.4780 Odour: Turpentine-like Melting Point: -61° Boiling Point: 164-165° Beilstein Reference: 2038282 Merck Reference: 14,7446 Optical Rotation: -21° (neat) UN No.: 2319 MDL No.: MFCD00001345

Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 164…165 °C
Melting Pt: –61 °C
Density: 0.860
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001345
CAS Number: 18172-67-3
EINECS: 242-060-2
UN: 2319
ADR: 3 III
Merck Index: 12,07600

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR