Dexamethasone ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion

A17590.06 A17590.03 A17590.14
A17590.06EA 214 GBP
A17590.06 A17590.03 A17590.14
Dexamethasone ≥98%
Dexamethasone

Purity: 98% CAS No.: 50-02-2 Molecular Formula: C22H29FO5 Formula Weight: 392.47 Melting Point: ca 260° dec. Beilstein Reference: 2066651 Merck Reference: 14,2943 Optical Rotation: +75° (c=1 in doxane) MDL No.: MFCD00064136


Anti-inflammatory glucocorticoid

Formula: C₂₂H₂₉FO₅
Melting Pt: ∼ 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064136
CAS Number: 50-02-2
EINECS: 200-003-9
Merck Index: 14,2943

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR