Chlorphenesin 99

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A17386.14 A17386.22 A17386.36
A17386.14EA 27.4 GBP
A17386.14 A17386.22 A17386.36
Chlorphenesin 99
Chlorphenesin

Purity: 99% CAS No.: 104-29-0 Molecular Formula: C9H11ClO3 Formula Weight: 202.64 Melting Point: 78-82° MDL No.: MFCD00021990

Formula: C₉H₁₁ClO₃
MW: 202.64 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD00021990
CAS Number: 104-29-0
EINECS: 203-192-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR