Sodium metabisulphite ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Disodium disulphite, di-Sodium disulphite, Sodium pyrosulphite, Disodiumdisulphite, Sodium disulphite

A17351.0C A17351.0E A17351.36
A17351.0CEA 105 GBP
A17351.0C A17351.0E A17351.36
Sodium metabisulphite ≥97%
Sodium metabisulphite

Purity: 97% CAS No.: 7681-57-4 Molecular Formula: Na2O5S2 Formula Weight: 190.10 Odour: Like rotten eggs pH: 3.5-5.0 Melting Point: 300° Merck Reference: 14,8638 MDL No.: MFCD00167602

Formula: Na₂S₂O₅
Melting Pt: >300 °C
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 14,8638
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR