O-Dianisidine dihydrochloride ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 3,3'-Dimethoxybenzidine dihydrochloride

A17175.03 A17175.06
A17175.03EA 20.6 GBP
A17175.03 A17175.06
O-Dianisidine dihydrochloride ≥99%
o-Dianisidine dihydrochloride

Purity: 99% CAS No.: 20325-40-0 Molecular Formula: C14H16N2O2.2HCl Formula Weight: 317.21 Melting Point: 268° dec. Beilstein Reference: 3917996

Formula: C₁₄H₁₈Cl₂N₂O₂
Melting Pt: 268 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012488
CAS Number: 20325-40-0
EINECS: 243-737-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR