Xanthotoxin 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

A17108.03 A17108.06 A17108.14
A17108.03EA 42.7 GBP
A17108.03 A17108.06 A17108.14
Xanthotoxin 99%
Xanthotoxin

Purity: 99% CAS No.: 298-81-7 Molecular Formula: C12H8O4 Formula Weight: 216.20 Melting Point: 146-150° Beilstein Reference: 196453 MDL No.: MFCD00005009

Formula: C₁₂H₈O₄
Melting Pt: 146…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005009
CAS Number: 298-81-7
EINECS: 206-066-9
Merck Index: 14,5988

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR