(1-Bromoethyl)benzene ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 1-Bromo-1-phenylethane, α-Methylbenzyl bromide

A16839.14 A16839.22 A16839.36
A16839.14EA 39.3 GBP
A16839.14 A16839.22 A16839.36
(1-Bromoethyl)benzene ≥97%
1-Bromo-1-phenylethane

Purity: 97% CAS No.: 585-71-7 Molecular Formula: C8H9Br Formula Weight: 185.06 Flash Point: 81°(177°F) Refractive Index: 1.5600 Melting Point: -65° Boiling Point: 200° Beilstein Reference: 507210 MDL No.: MFCD00000139

Formula: C₆H₅CHBrCH₃
Boiling Pt: 200 °C
Melting Pt: –65 °C
Density: 1.356
Flash Pt: 81 °C (177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000139
CAS Number: 585-71-7
EINECS: 209-560-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f329f7855ae4dffcd097f49d6d504b1ec165e692&vid=6d612184698aeb1969948bdeb536d2ee39ec9551&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR