4,4'-Diaminodiphenyl ether 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: p-Aminophenyl ether, 4,4'-Oxydianiline, 4,4-Oxydianiline, Bis(4-aminophenyl) ether, 4-Aminophenyl ether

A16815.22 A16815.18 A16815.36
A16815.22EA 57.1 GBP
A16815.22 A16815.18 A16815.36
4,4'-Diaminodiphenyl ether 98%
4,4'-Diaminodiphenyl ether

Purity: 98% CAS No.: 101-80-4 Molecular Formula: C12H12N2O Formula Weight: 200.24 Flash Point: 218°(424°F) Melting Point: 189-192° Boiling Point: 190°/0.1mm Beilstein Reference: 475735 UN No.: 3077 MDL No.: MFCD00007863

Formula: C₁₂H₁₂N₂O
Boiling Pt: 190 °C (0.1 mmHg)
Melting Pt: 189…192 °C
Flash Pt: 218 °C (424 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007863
CAS Number: 101-80-4
EINECS: 202-977-0
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR