1,1'-Ferrocenedicarboxylic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

A16691.03 A16691.06 A16691.14
A16691.03EA 50.6 GBP
A16691.03 A16691.06 A16691.14
1,1'-Ferrocenedicarboxylic acid ≥98%
1,1'-Ferrocenedicarboxylic acid

Purity: 98% CAS No.: 1293-87-4 Molecular Formula: C12H10FeO4 Formula Weight: 274.06 Odour: Odourless Melting Point: 300° MDL No.: MFCD00001423

Formula: C₁₂H₁₀FeO₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00001423
CAS Number: 1293-87-4
EINECS: 215-068-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR