Calcium nitrate tetrahydrate 97+% (by complexometric analysis)

Supplier: Thermo Fisher Scientific
Ratings:
Avg. Ratings: 1.0 (1 Review)

Total Ratings: 1
Avg. Ratings: out of 5

Danger

A16645.0E A16645.22 A16645.36
A16645.0EEA 93.7 GBP
A16645.0E A16645.22 A16645.36
Calcium nitrate tetrahydrate 97+% (by complexometric analysis)
Calcium nitrate tetrahydrate

Purity: 99% CAS No.: 13477-34-4 Molecular Formula: Ca(NO3)2.4H2O Formula Weight: 236.15 (164.09anhy) Odour: Odourless Melting Point: 43-45° Boiling Point: 132° dec. Merck Reference: 14,1682 UN No.: 1454 MDL No.: MFCD00149604

Formula: Ca(NO₃)₂·4H₂O
Boiling Pt: 132 °C (dec.)
Melting Pt: 43…45 °C
Density: 1.820
Storage Temperature: Ambient
MDL Number: MFCD00149604
CAS Number: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1,III
Merck Index: 14,1682

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR