3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Benzophenone-3,3',4,4'-tetracarboxylic dianhydride

A16529.22 A16529.36 A16529.A4
A16529.22EA 27.9 GBP
A16529.22 A16529.36 A16529.A4
3,3',4,4'-Benzophenonetetracarboxylic dianhydride ≥97%
3,3',4,4'-Benzophenonetetracarboxylic dianhydride

Purity: 97+% CAS No.: 2421-28-5 Molecular Formula: C17H6O7 Formula Weight: 322.23 Flash Point: 324°(615°F) Melting Point: 220-226° Boiling Point: 320°/5mm Beilstein Reference: 761777 MDL No.: MFCD00005923

Formula: C₁₇H₆O₇
Boiling Pt: 320 °C (5 mmHg)
Melting Pt: 220…226 °C
Density: 1.570
Flash Pt: 324 °C (615 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005923
CAS Number: 2421-28-5
EINECS: 219-348-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR